Learn More
Medchemexpress LLC Inixaciclib | 2370913-42-9 | 99.9% | C26H30F2N6O | 5 MG

Supplier: Medchemexpress LLC HY456615MG
Inixaciclib is a potent CDK inhibitor primarily utilized for anticancer research. This compound is known for its high purity and specific storage requirements to maintain its efficacy.
- Potent CDK inhibitor
- Used for anticancer research
- High purity of 99.9%
- Stable as powder at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.