Learn More
Medchemexpress LLC Inixaciclib | 2370913-42-9 | 99.9% | C26H30F2N6O | 50 MG

Supplier: Medchemexpress LLC HY4566150MG
Inixaciclib is a potent CDK inhibitor, which can be used for anticancer research.
- Purity of 99.93%
- Molecular weight of 480.55
- Solid appearance
- White to off-white color
- Ships at room temperature in continental US
- Stable as powder at -20°C for 3 years, 4°C for 2 years
- Stable in solvent at -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO at 11.9 mg/mL (24.76 mM)
- Soluble in 10% DMSO and 90% (20% SBE-β-CD in Saline) at ≥ 1.25 mg/mL (2.60 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.