Learn More
Glass Expansion Injector Adaptor for D-Torch
SDP
Supplier:  Glass Expansion 318083578
Encompass_Preferred
Injector Adaptor for D-Torch. Compatible with Glass Expansion D-Torches for Agilent 5000 SVDV/VDV/RV Series ICP-OES. The D-Torch Replacement O-ring Kit P/N 70-803-1500 includes all necessary O-rings for the D-Torch, including the Injector Adaptor. To help facilitate O-ring removal and installation, use the O-ring removal tool P/N 50-100-1636.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3123