Learn More
Glass Expansion Injector Adaptor for Semi-Demountable Torch
SDP
Supplier:  Glass Expansion 318084129
Encompass_Preferred
Injector Adaptor for Semi Demountable Torch. Compatible with Glass Expansion’s Semi Demountable Torch (NC2046974, 30-808-3853 and 30-808-4185) for Agilent 7700/7800/7900/8800 and 8900 ICP-MS, and Glass Expansion’s Semi Demountable Torch (30-808-4335) for Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3100