Learn More
Glass Expansion Injector Ferrule Tool - 6.0 mm
SDP
Supplier:  Glass Expansion 708030920
Encompass_Preferred
Injector Ferrule Tool - 6.0 mm. Provided with most Glass Expansion D-Torches to facilitate the removal and installation of the injector ferrule. ONLY compatible with Injector Ferrule 31-808-2962. Not compatible with Injector Ferrule (31-808-2897) for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES D-Torches . The injector ferrule is a consumable part that needs to be replaced on average about every 6 months. This tool should be used to remove and install the injector ferrule. Replacing the injector ferrule on a routine basis ensures a proper injector seal for optimum analytical performance.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3077