Learn More
Trajan Scientific INJECTOR SAMPLE 10F-CTC-0.47
SDP
Supplier:  Trajan Scientific 002980
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Not gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 50 mm, Connection Type: Fixed
Catalog No. 50-253-0360