Learn More
Trajan Scientific INJECTOR SAMPLE 10R-CTC-0.47
SDP
Supplier:  Trajan Scientific 002982
Encompass_Preferred
SGE PAL Autosampler Syringes with Removable 50mm Cone Tip Needle, Description: Not gas tight, Diameter Outer Needle: 0.47 mm, Volume: 10 uL, Needle Gauge: 26 G, Length Needle: 50 mm, Connection Type: Removable
Catalog No. 50-253-0358