Learn More
Instech Injectors to manually access the ports in Instech VAH95AB Vascular Access Harnesses, VAB95BS buttons and SIP22/4 injection ports. Needlefree. 50 sterile pouches of 5. Replaces older SN22 needles.
SDP
Supplier:  Instech VAH6M
Encompass
The Instech VAH6M is a specialized injector designed for accessing rat Vascular Access Harnesses, enabling safe manual flushing, dosing, and sampling through the septum without causing damage. Featuring integrated needle‑stick protection and engineered for compatibility with standard rat VAH ports, it supports reliable aseptic access during infusion or blood‑sampling protocols. The VAH6M is supplied sterile and is commonly paired with 3Fr rat catheters for optimal septum alignment.
Features
  • Injector for rat Vascular Access Harness systems
  • Designed for safe septum access without damage
  • Integrated needle‑stick protection
  • Compatible with one‑ and two‑channel rat VAH models
  • Supports flushing, dosing, and sampling
  • Recommended for use with 3Fr catheter systems
  • Sterile; available in bulk pack formats
Catalog No. 50-890-172