Learn More
Essendant INKMAINTENANCE BOXC9345
Supplier:  Essendant EPSC12C934591
Encompass
The C9345 Ink Maintenance Box from Epson is designed for select Expression printers This user-replaceable tank will keep your printer parts clean while maintaining optimal performance It is responsible for storing the ink that gets flushed from the system during printhead cleaning When capacity is getting low or the box needs to be changed your printer and software will alert you that maintenance tank needs replaced This box is easy to install Accessory Type- Ink Maintenance Box For Use With- Epson EcoTank Photo ET-8500 ET-8550 EcoTank Pro ET-16600 WorkForce EC-C7000 ST-C8000 ST-C8090 WorkForce Pro WF-7310 WF-7820 WF-7840
Catalog No. 50-000-32686