Learn More
Grainger Inline pH Kit CPVC BNC Flat Tip
Supplier:  Grainger 16Z024
Encompass_Preferred
The SENSOREX S6600 10 BNC Inline pH Kit is a pH measurement kit designed for inline pH monitoring in industrial water applications. It includes a S6600 pH sensor with 10ft BNC connector cable, S651 pH controller with alarm relay, flow cell fitting, and buffer solutions for calibration. The kit provides continuous pH measurement from 0-14 pH range with ±0.1 pH accuracy. The pH sensor has a replaceable cartridge design and the controller features automatic temperature compensation. This industrial pH measurement system is suitable for monitoring pH in water treatment, food processing, chemical processing, and other water-based applications.
Catalog No. 50-276-5152