Learn More
Med Vet International Inoculating Loop with Handle, 3mm x 2, J0549C
Supplier:  Med Vet International JOR549C
Encompass_Preferred
Inoculating loops are a necessary part of doing any sort of bacterial culturing. These Nichrome wire loops are attached to an anodized aluminum handle. The wire loops are durable plus rapidly cool following heat or flame sterilization. The loop is a 3mm diameter or 26g wire. Each.
Catalog No. 50-005-82476