Learn More
Medchemexpress LLC Inolitazone dihydrochloride | 223132-38-5 | 98.9% | 575.51 g/mol | C27H28Cl2N4O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14792B10MG
Encompass_Preferred
Inolitazone dihydrochloride is a research-grade, high-affinity peroxisome proliferator-activated receptor gamma (PPARγ) agonist. It is PPARγ-dependent for its biological activity and has demonstrated potent growth-inhibitory effects (reported IC50 ≈ 0.8 nM). The compound is used in cell proliferation and combination therapy studies and is supplied for research use only.
  • High-affinity PPARγ agonist with potent activity.
  • Demonstrated growth inhibition with IC50 ≈ 0.8 nM.
  • Upregulates p21 (WAF1/CIP1), useful in cell cycle studies.
  • Suitable for cell proliferation and combination therapy research.
  • Provided at high purity (≈98.9%) suitable for analytical assays.
  • Available in multiple small pack sizes for laboratory use.
Catalog No. 50-002-86085