Learn More
Medchemexpress LLC Inophyllum E | 17312-31-1 | 402.44 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN136451MG
Encompass_Preferred
Inophyllum E is a natural product that has shown antiproliferative activity against human HL60 and HeLa cells, and cytotoxicity in Molt-4 cells.
  • Antiproliferative activity against human HL60 cells (IC50 > 60 μM)
  • Antiproliferative activity against human HeLa cells (IC50 = 21.6 μM)
  • Cytotoxicity in Molt-4 cells (IC50 = 6.2 μM)
  • For research use only
Catalog No. 50-004-16126