Learn More
Cayman Chemical InosIn 5trIphosphATsodI 100mg
SDP
Supplier:  Cayman Chemical 23063100 MG
Encompass_Preferred
A nucleoside triphosphate; alternative substrate for ATPases and GTPases; has been used to study activation and binding kinetics of nucleoside interaction with various ATPases and GTPases
Catalog No. 50-299-5644