Learn More
Medchemexpress LLC Inosine-2,8-d2 | 697807-01-5 | 99.7% | 270.24 | C10H10D2N4O5 | 10 MG

Supplier: Medchemexpress LLC HYN0092S10MG
Inosine-2,8-d2 is a deuterium-labeled inosine supplied as a solid reference standard for analytical and research use, including mass spectrometry and quantitative assays.
- Deuterated inosine for isotopic labeling and internal standards.
- High purity: 99.7%.
- Molecular weight 270.24 g/mol and chemical formula C10H10D2N4O5.
- Solid form suitable for laboratory handling.
- Recommended storage under nitrogen at -20°C; in solution store at -80°C for up to 6 months.
- Packaged as 10 MG for small-scale analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.