Learn More
Apexbio Technology LLC Inosine 58-63-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B36775.1
Encompass_Preferred
Inosine (58-63-9) is an endogenous purine nucleoside It participates in purine catabolic and salvage pathways facilitating nucleotide synthesis and uric acid formation Inosine is structurally composed of hypoxanthine linked to a ribose sugar moiety It is also present in specific tRNA anticodons influencing translational processes Inosine exerts its biological activity by serving as an intermediate in purine metabolism and modulating nucleotide recycling and tRNA modifications In biochemical and cell biology studies inosine is utilized as a tool compound to investigate purine metabolism nucleotide recycling pathways tRNA modifications receptor-mediated signaling and immune regulation in vitro Reported IC50 values may vary depending on biological models and experimental conditions applied Based on these pharmacological properties inosine holds research potential in studies of purine metabolism nucleotide synthesis RNA biology and immune regulation
Catalog No. 50-000-89662