Learn More
Medchemexpress LLC Inosine pranobex | 36703-88-5 | MFCD05662374 | 98.1% | 1115.23 | C52H78N10O17 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10780110MM 1ML
Encompass_Preferred
Inosine pranobex is an orally active immunomodulator with broad-spectrum antiviral activity commonly used in research. It is supplied in both solid and DMSO solution formats, and has defined solubility and storage profiles suitable for in vitro and formulated in vivo studies.
  • Orally active immunomodulator with antiviral activity.
  • High purity (98.1%) for reproducible experimental results.
  • Available as solid and as a 10 mM solution in DMSO for convenient dosing.
  • Soluble in DMSO (≈62.5 mg/mL) with documented in vivo formulation options.
  • Stable when stored under recommended powder and solvent conditions.
Catalog No. 50-002-44486