Learn More
Medchemexpress LLC HY-107801 500mg Medchemexpress, Inosine pranobex CAS:36703-88-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107801500MG
Encompass_Preferred
Medchemexpress, HY-107801 500mg Inosine pranobex CAS:36703-88-5 Inosine pranobex is a potent, broad-spectrum antiviral compound for HIV infection. Inosine pranobex is an immunopotentiator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0721