Learn More
MSC Insize Dial Test Indicator Holder, 5.354 in Length, 3/8 in Stem Diameter
Supplier: MSC 12918033
This Dial Test Indicator Holder provides a stable platform for dial test indicators, designed for precise measurement applications. It features specific dimensions and accessories to ensure compatibility and ease of use in various inspection and measurement tasks.
- Overall length: 5.354 in
- 3/8 in Diameter holding stem
- Stem hole diameters: 5/32 in and 1/4 in
- Dovetail groove
- Supplied with hex key
- For use with dial test indicators