Learn More
MSC Value Collection Insulated Shipping Box, 19-1/2 in L, 1 kit
Supplier:  MSC 39559331
Encompass
This insulated shipping kit is designed to maintain the insulation and integrity of temperature-sensitive products. It is economical, lightweight, reusable, and recyclable, featuring a seamless molded EPS foam body with a tight-fitting lid and a durable white corrugated carton.
  • Economical, lightweight, reusable, and recyclable
  • Maintains insulation and integrity of temperature-sensitive products
  • Seamless molded EPS foam body with tight-fitting lid
  • Includes 200#/ECT-32 white corrugated carton
  • Cartons pre-printed with 'Rush, Perishable,' and 'This Side Up' shipping instructions
  • Thick molded foam container protects contents from crushing and breakage
Catalog No. 50-191-6396