Learn More
MSC Polar Tech Insulated Shipping Box, 12x10x7 in, 1 ea
Supplier:  MSC 39559265
Encompass
This insulated shipping box is designed to maintain the integrity of temperature-sensitive products. It features a seamless molded EPS foam body with a tight-fitting lid, enclosed within a durable white corrugated carton. This economical and lightweight kit is reusable and recyclable.
  • Seamless molded EPS foam body with tight-fitting lid
  • Constructed with 1.5 in polystyrene foam for superior insulation
  • Includes 200#/ECT-32 white corrugated carton
  • Carton pre-printed with 'Rush,' 'Perishable,' and 'This Side Up' shipping instructions
  • Reusable and recyclable materials
  • Rectangle container shape
  • White color
Catalog No. 50-191-6392