Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Inspection Microscope zoom 10-40x
SDP
Supplier:  Electron Microscopy Sciences 700011
Encompass_Preferred
Optional binocular inspection microscope for EMS7000 Vibrating Microtomes has 10x-40x magnification, a zoom of 1x-4x and a working distance of 80mm.
Catalog No. 50-949-911