Learn More
MSC LPS Instant Super Degreaser 2.0, 55 gal, Drum, Orange Clean
Supplier:  MSC 38934147
Encompass
LPS Instant Super Degreaser 2.0 is a heavy-duty, non-flammable degreaser that rapidly evaporates, leaving no residue. It is designed to remove oil, grease, dirt, and tar effectively, enhancing safety and productivity in various applications.
  • Can be used on or near energized equipment as it is non-flammable and non-conductive.
  • Fast evaporation quickens clean-up time.
  • Requires no rinsing or wiping prior to the next production step.
  • Removes oil, grease, dirt, and tar with less cleaner.
  • Improved health and safety profile.
  • Reduces time, labor, and equipment downtime.
Catalog No. 50-283-6009
Add to Cart
Request a Quote