Learn More
Sklar Instruments Instrument Tray, Sklar, 20-3/4 in. x 12-3/4 in. x 6 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. EA/1.
SDP
Supplier:  Sklar Instruments 101948
Encompass_Preferred
Instrument Tray, Sklar, 20-3/4 in. x 12-3/4 in. x 6 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 20-3/4 inches x 12-3/4 inches x 6 inches.
Catalog No. 50-143-7153