Learn More
Genetex Inc Insulin Monoclonal Antibody [HMV308] HistoMAX™ (GTX639932) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639932
Encompass_Preferred
The Insulin antibody [HMV308] HistoMAX™ (GTX639932) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human pancreatic neuroendocrine tumor (PNET, insulinoma) tissue using GTX639932 Insulin antibody [HMV308] HistoMAX. Strong Insulin positivity of most tumor cells.
Catalog No. NC3669302