Learn More
Medchemexpress LLC Insulin (cattle) | 11070-73-8 | 98.4% | 5800 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1156
Encompass_Preferred
Insulin (cattle) is a two-chain polypeptide hormone produced by the pancreatic β cells. It is commonly used as a growth supplement in cell culturing. In vitro, the α and β chains are joined by two interchain disulfide bonds, and the α chain contains an intrachain disulfide bond. This hormone regulates glucose uptake into muscle and fat cells by recruiting membrane glucose transporter Glut-4 to the cell surface. It has been effectively utilized in cell cultures at concentrations ranging from 1 to 10 μg/mL.
  • Two-chain polypeptide hormone
  • Produced by pancreatic β cells
  • Used as a growth supplement in cell culturing
  • Regulates glucose uptake into muscle and fat cells
  • Recruits membrane glucose transporter Glut-4 to the cell surface
  • Effective for cell culturing at 1 to 10 μg/mL concentration
Catalog No. 50-004-26658