Learn More
Medchemexpress LLC Insulin (human), 21A-glycine-30Ba-L-arginine-30Bb-L-arginine- | 160337-95-1 | ≥95.0% | 6062.89 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108719100U
Encompass_Preferred
Insulin glargine is a long-acting insulin analog that lowers blood sugar and can be used in diabetes research. High doses of Insulin glargine may also promote the proliferation of bladder cancer cells.
  • Long-acting insulin analog
  • Lowers blood sugar
  • Promotes proliferation of bladder cancer cells at high doses
Catalog No. 50-003-46323