Learn More
BioVendor, LLC Insulin Human, E. coli Recombinant Protein, 1 g
SDP
Supplier:  BioVendor, LLC RP1762701000
Encompass
Insulin Human, E. coli Recombinant Protein, 1 g. Research use only. Recombinant human insulin is commonly used in the growth of mammalian cell lines. Human Insulin recombinantly produced in E. coli is a two chain, non-glycosylated polypeptide chain containing 51 amino acids and having a molecular mass of 5807 Dalton. Sterile filtered white lyophilized (freeze-dried) powder. Lyophilized from a concentrated (1 mg/ml) solution with no additives. Purity > 98%. Biological activity: 28 units/mg. Summary: Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver.
Catalog No. 50-206-9280