Learn More
Medchemexpress LLC Insulin Human P.pa 2ug
SDP
Supplier:  Medchemexpress LLC HYP732432UG
Encompass_Preferred
Insulin is an important hormone in glucose regulation reducing blood glucose concentrations by increasing cellular permeability to simple sugars amino acids and fatty acids Its multiple effects include accelerating glycolysis promoting pentose phosphate cycle and promoting hepatic glycogen synthesis Insulin Protein Human (P pastoris) is the recombinant human-derived Insulin protein expressed by P pastoris with tag free
Catalog No. 50-001-80288