Learn More
Medchemexpress LLC Insulin levels modulator | 1019254-94-4 | 98.4% | 421.52 | 25 MG

Supplier: Medchemexpress LLC 25MGHY112819
Insulin levels modulator is an insulin secretagogue that can be used for the study of a disease associated with insufficient insulin production, such as type 2 diabetes.
- An insulin secretagogue
- Used for the study of diseases associated with insufficient insulin production
- Suitable for type 2 diabetes research
- Purity of 98.38%
- Molecular weight: 421.52
- Appears as a white to off-white solid
- Store at 4°C, protect from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.