Learn More
Cusabio Technology LLC Rat Insulin-like growth factor 1IGF-1 ELISA Kit
Supplier:  Cusabio Technology LLC CSBE04582R
Encompass_Preferred
This Rat IGF1 ELISA Kit was designed for the quantitative measurement of Rat IGF1 protein in serum, plasma, cell culture supernates, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 0.156 ng/mL-10 ng/mL and the sensitivity is 0.151 ng/mL.
Catalog No. NC3654169