Learn More
STEMCELL Technologies Human Recombinant IGF-II, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78023.2
Encompass
Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways. IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development. It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova).
Catalog No. 50-226-9632