Learn More
Cusabio Technology LLC Fish Insulin-like growth factors 1,IGF-1 ELISA Kit
Supplier:  Cusabio Technology LLC CSBE12122FH
Encompass_Preferred
This Fish IGF-1 ELISA Kit was designed for the quantitative measurement of Fish IGF-1 protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 25 pg/mL-2000 pg/mL and the sensitivity is 31.25 pg/mL. One kit contains 96 wells.
Catalog No. NC2952942