Learn More
Medchemexpress LLC Insulin lispro | 133107-64-9 | 5807.63 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY1095301MG
Encompass_Preferred
Insulin lispro is a recombinant human insulin analogue and is one of three rapid-acting insulin analogues available. It can be used for the research of hyperglycaemia in diabetes mellitus.
  • For research use only.
  • Available in multiple sizes.
  • Estimated time of arrival for select sizes.
  • Controlled substance, not for sale in all territories.
Catalog No. 50-003-45317