Learn More
Alpco Ltd Rat High Range Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSRTHE01
Encompass_Preferred
The Rat High Range Insulin ELISA quantifies the concentration of insulin protein products in rat I and rat II proinsulin genes with as little as 5 µL of serum or plasma sample.
  • Only 5 µL of sample is sufficient for the assay
  • Superior sensitivity of 0.52 ng/mL
  • Dynamic range of 3-150 ng/mL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-594