Learn More
Medchemexpress LLC Insulin receptor (1142-1153), pTyr1150 | 134177-41-6 | 99.4% | 1702.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP54315MG
Encompass_Preferred
Insulin receptor (1142-1153), pTyr1150 is a biologically active peptide. It is used as an insulin receptor tyrosine kinase substrate.
  • Biologically active peptide
  • Used as an insulin receptor tyrosine kinase substrate
Catalog No. 50-003-98600