Learn More
Medchemexpress LLC Insulin Receptor Antibody (YA3666) | 100 UL

Supplier: Medchemexpress LLC HYP83969100UL
Insulin Receptor Antibody (YA3666) is a laboratory chemical designed for research use. It is supplied as a liquid and has predicted and observed molecular weights.
- Host: Mouse
- Reactivity: Human, Mouse, Monkey
- Clonality: YA3666
- SwissProt ID: P06213
- Gene ID: 3643
- Application: WB, ICC/IF, FC, ELISA
- Dilution ratio: WB: 1:500-1:2000; ICC/IF: 1:50-1:200; FC: 1:200-1:400; ELISA: 1:10000
- Formulation: Supplied in PBS with 0.05% sodium azide
- Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.