Learn More
Supply Solutions Insulin solution human, 11061-68-0, MFCD00131380, 5mL
Supplier:  Supply Solutions SIGMA I92785ML
Encompass_Preferred
Chemically defined, recombinant, expressed in Saccharomyces cerevisiae, sterile-filtered, BioXtra, Insulin solution human is recommended for use in cell culture applications at 0.5 to 1mL per liter of medium.
Catalog No. 50-161-3342