Learn More
Grainger InsulMultitapCon PVC 4 AWG 500 kcmil
Supplier:  Grainger 14V970
Encompass_Preferred
The Grainger Insulated Multitap Wire Connectors are designed for connecting multiple 4/0 AWG copper or aluminum wires. They feature a split bolt design with an insulated cover for safety. The connectors accommodate wire sizes from #14-4/0 AWG. They are UL listed for 600V building wire applications. The connectors are available in 2-port, 3-port, 4-port, and 5-port options. They provide a secure mechanical connection without soldering or taping. The insulated cover prevents accidental contact.
Catalog No. 50-274-9526