Learn More
Grainger InsulMultitapCon PVC 6 AWG 250 kcmil
Supplier:  Grainger 14V968
Encompass_Preferred
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single source. It is rated for 600 volts and can accommodate wire sizes from 12 to 6 AWG. The connector has 4 ports and is made from UV resistant polycarbonate. It can be used for a variety of indoor and outdoor applications including landscape lighting, pools, spas, and signage. The connector measures 4.5 x 2.75 x 1.5 inches and has an operating temperature range of -40°F to 221°F. It is cULus listed for safety.
Catalog No. 50-274-8787