Learn More
Grainger InsulMultitapCon PVC 6 AWG 3/0 AWG
Supplier:  Grainger 14V974
Encompass_Preferred
The Polaris InsulMultitapCon PVC 6 AWG is a 6 AWG insulated multitap connector made of PVC. It has a temperature rating of 105°C and is UL listed. This multitap connector accommodates 6 to 12 wires from #14 to #6 AWG. It features a clear cover for quick inspection and wire insertion holes for easy installation. The connector measures 2-1/4" H x 2-5/8" W x 1-3/8" D. This PVC multitap connector is ideal for splicing and tapping power and control wires.
Catalog No. 50-274-9562