Learn More
Medchemexpress LLC HY-12434 5mg Medchemexpress, INT-767 CAS:1000403-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124345MG
Encompass_Preferred
Medchemexpress, HY-12434 5mg INT-767 CAS:1000403-03-1 INT-767 is a dual farnesoid X receptor (FXR)/TGR5 agonist with mean EC50s of 30 and 630 nM, respectively[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9988