Learn More
Waters Corp WATERS CORPORATION Integrated Guard Holder, for Chromatography Systems, Quick-Release Mechanism
Supplier:  Waters Corp WAT046905
Encompass_Preferred
The Integrated Guard Holder is a high-quality accessory designed for use with chromatography systems. It is constructed from durable materials to ensure long-lasting performance and reliability. This essential tool is designed to hold guard cartridges, which protect the analytical column from contaminants, thereby extending its lifespan. It features a quick-release mechanism, allowing for efficient guard cartridge changes, which reduces downtime and increases productivity.
  • Designed for use with chromatography systems
  • Made with durable materials for long-lasting performance and reliability
  • Holds guard cartridges
  • Protects the analytical column from contaminants
  • Extends the lifespan of the analytical column
  • Easy-to-use features
  • Quick-release mechanism for efficient guard cartridge changes
  • Reduces downtime and increases productivity
Catalog No. 50-785-636