Learn More
ABclonal Technology Integrin alpha 1 ITGA1/CD49a
SDP
Supplier:  ABclonal Technology A2270720UL
Encompass_Preferred
This gene encodes the alpha 1 subunit of integrin receptors This protein heterodimerizes with the beta 1 subunit to form a cell-surface receptor for collagen and laminin The heterodimeric receptor is involved in cell-cell adhesion and may play a role in inflammation and fibrosis The alpha 1 subunit contains an inserted (I) von Willebrand factor type I domain which is thought to be involved in collagen binding
Catalog No. 50-000-07671