Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 8 protein, mouse | ≥95.0% | 20UG
SDP

Supplier:  Medchemexpress LLC HYP70076320UG

Encompass_Preferred

Integrin alpha V beta 8 protein (mouse) is a recombinant heterodimer expressed in HEK293 cells with a C-terminal His tag, intended for research use in studies of integrin signaling, ligand binding, and cell adhesion. The product is supplied as a purified protein in defined sizes and is characterized by a reported purity and molecular weight range suitable for biochemical and cell-based assays.

  • Recombinant mouse integrin heterodimer suitable for functional assays.
  • Produced in HEK293 expression system to preserve mammalian modifications.
  • His tag facilitates purification and detection in biochemical workflows.
  • High purity supports reliable assay results (≥95.0%).
  • Supplied in defined sizes for flexible experimental needs.
  • Molecular weight consistent with alpha and beta subunits (80-140 kDa).

Catalog No. 50-001-77247


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.