Learn More
Sino Biological Recombinant Human Integrin alpha V beta 3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological CT098H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Val992) of human ITGAV (P06756) was fused with a his tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asp718) of human ITGB3 (P05106) constructed the plasmid 2. The two plasmids were co-expressed and the human ITGAV&ITGB3 heterodimer was purified.
Catalog No. 50-211-5530