Learn More
ECM Bioscience Integrin β4 Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience IK6270
Encompass_Preferred
The Integrin β4 phospho-regulation antibody sampler kit can be used to detect phosphorylation of Integrin β4 at Tyr-1494 and Tyr-1526. The kit also includes an antibody to examine total Integrin β4 expression levels and secondary reagents for rabbit polyclonal and mouse monoclonal antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8728