Learn More
Bioss Integrin L - Extracellular re
SDP
Supplier:  Bioss BSM70524M
Encompass_Preferred
Integrin L (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M594Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66384