Learn More
Medchemexpress LLC Integrin Linked ILK Antibody (YA3111) | 10 UL

Supplier: Medchemexpress LLC HYP8336610UL
Integrin Linked ILK Antibody (YA3111) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Integrin Linked ILK.
- Host: Rabbit
- Reactivity: Human, Mouse, Rat
- Conjugation: Non-conjugated
- SwissProt ID: Q13418
- Research field: Signal transduction
- Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles
- Appearance: Liquid
- Shipping: Shipping with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.