Learn More
LICOR Intercept T20 Protein-Free Antibody Diluent (3x500mL)
Supplier:  LICOR 92785003
Encompass_Preferred
Intercept T20 (TBS) Protein-Free Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (TBS) Protein-Free Blocking Buffer in Tris-buffered saline (TBS) preformulated with Tween 20. This diluent can be used where animal-based products are prohibited. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (TBS) Protein-Free Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.
Catalog No. NC2216062